Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166461.1 | NP_001159933.1 | 2 | 390 | ||
NM_001166462.1 | NP_001159934.1 | 3 | 539 | |||
NM_002436.3 | NP_002427.1 | 2 | 390 | |||
XM_011531167.1 | XP_011529469.1 | 2 | 384 | |||
GenBank mRNA | AK223396.1 | - | 2 | 370 | ||
AK290246.1 | - | 2 | 285 | |||
AK303111.1 | - | 2 | 205 | |||
AK312296.1 | - | 2 | 261 | |||
AK315957.1 | - | 3 | 388 | |||
AY423731.1 | - | 2 | 262 | |||
AY634686.1 | - | 2 | 260 | |||
BC002392.2 | - | 2 | 266 | 2820598 | ||
DA074686.1 | - | 3 | 539 | |||
M64925.1 | - | 2 | 339 |