Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153448.3 | NP_703149.1 | 3 | 597 | ||
GenBank mRNA | AK097704.1 | - | 3 | 612 | ||
AY114148.1 | - | 3 | 571 | |||
BC042633.1 | - | 3 | 558 | 5170565 | ||
BC053599.1 | - | 3 | 586 | 5742902 |