Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278682.1 | NP_001265611.1 | 3 | 1050 | ||
NM_001278684.1 | NP_001265613.1 | 4 | 1407 | |||
NM_001278686.1 | NP_001265615.1 | 4 | 1073 | |||
NM_003244.3 | NP_003235.1 | 3 | 1140 | |||
NM_170695.3 | NP_733796.2 | 3 | 1784 | |||
NM_173207.2 | NP_775299.1 | 3 | 1018 | |||
NM_173208.2 | NP_775300.1 | 4 | 1410 | |||
NM_173209.2 | NP_775301.1 | 3 | 1226 | |||
NM_173210.2 | NP_775302.1 | 3 | 1177 | |||
NM_173211.1 | NP_775303.1 | 3 | 889 | |||
NM_174886.2 | NP_777480.1 | 4 | 1207 | |||
XM_011525735.2 | XP_011524037.1 | 3 | 1386 | |||
XM_017025958.1 | XP_016881447.1 | 4 | 1471 | |||
XM_017025959.1 | XP_016881448.1 | 3 | 1256 | |||
GenBank mRNA | AK026766.1 | - | 2 | 1746 | ||
AK291112.1 | - | 3 | 974 | |||
BC000814.1 | - | 3 | 906 | 3453337 | ||
BC031268.1 | - | 3 | 1512 | 5284849 | ||
X89750.1 | - | 3 | 1133 |