Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198479.2 | NP_940881.2 | 2 | 926 | ||
GenBank mRNA | AK097640.1 | - | 2 | 926 | ||
BC137501.1 | - | 2 | 880 | 9021501 | ||
BC141863.1 | - | 2 | 868 | 40037835 | ||
BC144673.1 | - | 3 | 850 | 9053205 | ||
DQ340180.1 | - | 4 | 1146 |