Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035521.2 | NP_001030598.1 | 11, 12 | 1915 | ||
NM_001318909.1 | NP_001305838.1 | 11, 12 | 2025 | |||
NM_001521.3 | NP_001512.1 | 12, 13 | 1991 | |||
XM_005264272.4 | XP_005264329.1 | 12, 13 | 2199 | |||
XM_011532801.2 | XP_011531103.1 | 12, 13 | 2338 | |||
GenBank mRNA | AK307656.1 | - | 2, 3 | 121 | ||
BC000212.2 | - | 12, 13 | 1735 | 3508353 | ||
BC020981.1 | - | 10, 11 | 1624 | 3849379 | ||
BT009799.1 | - | 10, 11 | 1595 | |||
BX647092.1 | - | 11, 12 | 2025 | |||
D13636.1 | - | 10, 11 | 1634 |