Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323281.1 | NP_001310210.1 | 1 | 415 | ||
NM_001323282.1 | NP_001310211.1 | 1 | 415 | |||
NM_002895.4 | NP_002886.2 | 1 | 415 | |||
NM_183404.3 | NP_899662.1 | 1 | 415 | |||
XM_006723842.2 | XP_006723905.1 | 1 | 191 | |||
XM_011528955.2 | XP_011527257.1 | 1 | 191 | |||
XM_011528958.2 | XP_011527260.1 | 1 | 191 | |||
XM_017027992.1 | XP_016883481.1 | 1 | 229 | |||
GenBank mRNA | AK290380.1 | - | 1 | 191 | ||
AK308096.1 | - | 1 | 175 | |||
AK310870.1 | - | 1 | 200 | |||
BC032247.1 | - | 1 | 198 | 4521266 | ||
DC375054.1 | - | 1 | 417 | |||
L14812.1 | - | 1 | 196 |