Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003036.3 | NP_003027.1 | 7 | 3223 | ||
XM_005244775.3 | XP_005244832.1 | 7 | 3199 | |||
XM_005244776.4 | XP_005244833.1 | 7 | 6342 | |||
XM_017002128.1 | XP_016857617.1 | 7 | 3006 | |||
GenBank mRNA | AK309515.1 | - | 1 | 695 | ||
X15218.1 | - | 7 | 3222 |