Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002354.2 | NP_002345.2 | 7 | 1030 | ||
GenBank mRNA | AK026585.1 | - | 7 | 867 | ||
BC014785.1 | - | 7 | 854 | 3861826 | ||
CR542259.1 | - | 7 | 672 | |||
CR542283.1 | - | 7 | 672 | |||
M26481.1 | - | 7 | 831 | |||
M32306.1 | - | 7 | 693 | |||
M32325.1 | - | 7 | 826 | |||
M33011.1 | - | 7 | 779 | |||
X14758.1 | - | 7 | 826 |