Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003884.4 | NP_003875.3 | 2 | 832 | ||
XM_005265528.4 | XP_005265585.1 | 2 | 457 | |||
XM_017007423.1 | XP_016862912.1 | 3 | 287 | |||
XM_017007424.1 | XP_016862913.1 | 2 | 383 | |||
GenBank mRNA | BC060823.1 | - | 2 | 437 | 30336561 | |
BC070075.1 | - | 2 | 502 | 30333414 | ||
U57317.2 | - | 2 | 844 |