Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025081.1 | NP_001020252.1 | 7 | 1220 | ||
NM_001025090.1 | NP_001020261.1 | 6 | 1142 | |||
NM_001025092.1 | NP_001020263.1 | 5 | 1109 | |||
NM_001025101.1 | NP_001020272.1 | 9 | 1714 | |||
NM_002385.2 | NP_002376.1 | 6 | 1187 | |||
GenBank mRNA | AK094611.1 | - | 1 | 1306 | ||
AK095121.1 | - | 1 | 2098 | |||
AK098402.1 | - | 6 | 1098 | |||
AK122594.1 | - | 2 | 1665 | |||
AK123433.1 | - | 2 | 3762 | |||
AK124830.1 | - | 2 | 2439 | |||
AK126858.1 | - | 1 | 4252 | |||
AK128770.1 | - | 6 | 1101 | |||
AK128788.1 | - | 6 | 1098 | |||
AK289893.1 | - | 6 | 1142 | |||
AK293922.1 | - | 5 | 1137 | |||
BC008749.2 | - | 6 | 1080 | 3346521 | ||
BC068550.1 | - | 4 | 956 | |||
BC080654.1 | - | 5 | 1056 | 4859683 | ||
BC130034.1 | - | 1 | 117 | |||
CR627018.1 | - | 4 | 8868 | |||
M13577.1 | - | 6 | 1086 | |||
M30047.1 | - | 5 | 1050 | |||
M30515.1 | - | 8 | 1136 | |||
M30516.1 | - | 6 | 1106 |