Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MECOM SIRNA Assays ›
Gene Symbol:
MECOM
Gene Name:
MDS1 and EVI1 complex locus
Gene Aliases:
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location:
Chr.3: 169083499 - 169663781 on Build GRCh38
UniGene ID:
Hs.744090
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115629
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MECOM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105077.3
NP_001098547.3
15
3088
NM_001105078.3
NP_001098548.2
14
3079
NM_001163999.1
NP_001157471.1
13
2999
NM_001164000.1
NP_001157472.1
13
3928
NM_001205194.1
NP_001192123.1
14
3383
NM_004991.3
NP_004982.2
15
3721
NM_005241.3
NP_005232.2
14
3023
XM_005247213.3
XP_005247270.1
15
3730
XM_005247214.3
XP_005247271.1
14
3703
XM_005247215.3
XP_005247272.1
14
3699
XM_005247219.2
XP_005247276.1
14
3695
XM_005247220.2
XP_005247277.1
14
3036
XM_005247221.2
XP_005247278.1
14
3019
XM_005247223.2
XP_005247280.1
14
3692
XM_005247224.3
XP_005247281.1
15
2758
XM_005247225.3
XP_005247282.1
15
2754
XM_005247226.3
XP_005247283.1
14
2605
XM_011512546.2
XP_011510848.1
14
3483
XM_017005874.1
XP_016861363.1
14
4398
XM_017005875.1
XP_016861364.1
13
3006
XM_017005876.1
XP_016861365.1
13
3668
XM_017005877.1
XP_016861366.1
14
2730
XM_017005878.1
XP_016861367.1
13
2693
GenBank mRNA
AK025934.1
-
8
927
AK292865.1
-
14
3006
AK310509.1
-
12
2840
BC031019.1
-
7
1184
BC130520.1
-
15
3062
40146551
BC143951.1
-
15
3062
9052469
BC143952.1
-
13
2973
9052470
BX640908.1
-
13
3928
BX647613.1
-
13
3660
GQ352634.1
-
14
3287
X54989.1
-
14
3021
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
cell differentiation
histone lysine methylation
negative regulation of programmed cell death
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of JNK cascade
regulation of cell cycle
hematopoietic stem cell proliferation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0