Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 18, 19 | 2768 | ||
XM_005252051.2 | XP_005252108.1 | 18, 19 | 2765 | |||
XM_006717143.2 | XP_006717206.1 | 18, 19 | 2660 | |||
GenBank mRNA | AB014562.2 | - | 17, 18 | 2661 | ||
AK294861.1 | - | 6, 7 | 909 | |||
AL834263.1 | - | 18, 19 | 2619 | |||
BC028051.2 | - | 6, 7 | 879 | |||
BC064613.1 | - | 5, 6 | 341 | |||
BC152437.1 | - | 17, 18 | 2661 |