Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166343.1 | NP_001159815.1 | 7 | 1959 | ||
NM_002405.3 | NP_002396.2 | 8 | 2054 | |||
NR_029413.1 | 7 | 2005 | ||||
GenBank mRNA | BC094814.1 | - | 8 | 2015 | 5930073 | |
U94352.1 | - | 8 | 1963 |