Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 6, 7 | 935 | ||
NM_005426.2 | NP_005417.1 | 7, 8 | 1067 | |||
XM_011544268.1 | XP_011542570.1 | 5, 6 | 796 | |||
XM_011544269.2 | XP_011542571.1 | 3, 4 | 454 | |||
XM_017002220.1 | XP_016857709.1 | 7, 8 | 1076 | |||
GenBank mRNA | AJ318888.2 | - | 6, 7 | 880 | ||
AK294432.1 | - | 6, 7 | 787 | |||
BC040247.1 | - | 4, 5 | 411 | 4454266 | ||
BC046150.1 | - | 6, 7 | 878 | |||
BC058918.1 | - | 6, 7 | 893 | 4537206 | ||
U58334.1 | - | 7, 8 | 1012 |