Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206567.1 | NP_001193496.1 | 11 | 2506 | ||
NM_005531.2 | NP_005522.2 | 11 | 2506 | |||
XM_005245127.3 | XP_005245184.1 | 13 | 2991 | |||
XM_006711290.1 | XP_006711353.1 | 10 | 2338 | |||
XM_017001149.1 | XP_016856638.1 | 11 | 2655 | |||
GenBank mRNA | AB208989.1 | - | 11 | 2476 | ||
AF208043.1 | - | 10 | 3921 | |||
AK094968.1 | - | 9 | 1879 | |||
AK296228.1 | - | 11 | 2389 | |||
AK307957.1 | - | 11 | 2287 | |||
BC017059.1 | - | 11 | 2473 | 3914632 | ||
BM806453.1 | - | 2 | 285 | |||
M63838.1 | - | 11 | 2480 |