Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 2 | 469 | ||
NM_005933.3 | NP_005924.2 | 2 | 469 | |||
XM_006718839.3 | XP_006718902.2 | 2 | 461 | |||
XM_011542829.2 | XP_011541131.1 | 3 | 560 | |||
XM_011542830.2 | XP_011541132.1 | 3 | 560 | |||
XM_011542831.2 | XP_011541133.1 | 3 | 560 | |||
XM_011542833.2 | XP_011541135.1 | 3 | 560 | |||
GenBank mRNA | AF024540.1 | - | 88 | |||
AY187942.1 | - | 125 | ||||
AY187943.1 | - | 125 | ||||
AY187944.1 | - | 254 | ||||
AY187945.1 | - | 254 | ||||
D14540.1 | - | 2 | 469 | |||
L04284.1 | - | 2 | 446 | |||
L04731.1 | - | 2 | 260 |