Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115785
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
3
498
NM_001282691.1
NP_001269620.1
3
250
NM_006309.3
NP_006300.1
2
197
NM_017724.2
NP_060194.1
3
498
XM_005265539.2
XP_005265596.1
3
1197
XM_005265540.1
XP_005265597.1
4
480
XM_005265550.1
XP_005265607.1
3
465
XM_005265551.1
XP_005265608.1
3
465
XM_005265553.1
XP_005265610.1
3
465
XM_005265554.2
XP_005265611.1
3
1197
XM_005265556.1
XP_005265613.1
3
465
XM_006713385.1
XP_006713448.1
3
465
XM_006713386.1
XP_006713449.1
3
465
XM_006713387.1
XP_006713450.1
3
465
XM_006713388.1
XP_006713451.1
3
465
XM_006713389.1
XP_006713452.1
3
465
XM_006713390.1
XP_006713453.1
4
508
XM_006713392.1
XP_006713455.1
3
465
XM_006713393.1
XP_006713456.1
3
465
XM_006713394.1
XP_006713457.1
3
465
XM_006713395.1
XP_006713458.1
3
465
XM_006713396.1
XP_006713459.1
3
465
XM_006713397.1
XP_006713460.1
3
465
XM_006713400.1
XP_006713463.1
3
465
XM_006713404.1
XP_006713467.1
3
465
XM_011534217.1
XP_011532519.1
3
465
XM_011534218.1
XP_011532520.1
4
508
XM_011534219.1
XP_011532521.1
4
576
XM_011534221.1
XP_011532523.1
3
465
XM_011534222.1
XP_011532524.1
3
465
XM_011534229.1
XP_011532531.1
4
508
XM_017007465.1
XP_016862954.1
3
465
XM_017007466.1
XP_016862955.1
3
465
XM_017007467.1
XP_016862956.1
3
465
XM_017007468.1
XP_016862957.1
3
465
XM_017007469.1
XP_016862958.1
3
465
XM_017007470.1
XP_016862959.1
3
465
XM_017007471.1
XP_016862960.1
3
465
XM_017007472.1
XP_016862961.1
3
465
XM_017007473.1
XP_016862962.1
3
465
XM_017007474.1
XP_016862963.1
3
465
XM_017007475.1
XP_016862964.1
3
1197
XM_017007476.1
XP_016862965.1
4
508
XM_017007477.1
XP_016862966.1
3
1197
XM_017007478.1
XP_016862967.1
3
465
XM_017007479.1
XP_016862968.1
3
465
XM_017007480.1
XP_016862969.1
3
709
XM_017007481.1
XP_016862970.1
3
465
XM_017007482.1
XP_016862971.1
3
465
XM_017007483.1
XP_016862972.1
3
1197
XM_017007484.1
XP_016862973.1
4
508
GenBank mRNA
AF115509.1
-
2
156
AK000255.1
-
2
183
AK291514.1
-
3
475
AK302282.1
-
3
224
BC053668.1
-
3
455
6146978
DB033703.1
-
3
251
DC336346.1
-
3
500
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0