Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029976.2 | NP_001025147.2 | 4 | 2199 | ||
NM_006958.2 | NP_008889.2 | 3 | 2113 | |||
XM_005272341.2 | XP_005272398.1 | 3 | 2090 | |||
XM_005272342.2 | XP_005272399.1 | 3 | 2198 | |||
XM_011517298.1 | XP_011515600.1 | 3 | 2215 | |||
GenBank mRNA | AF244088.1 | - | 2 | 2024 | ||
AK127625.1 | - | 3 | 2113 | |||
AK226016.1 | - | 3 | 2083 | |||
AL834451.1 | - | 2 | 2030 | |||
BC010996.2 | - | 3 | 2047 | 3949487 | ||
DQ117529.1 | - | 4 | 2199 |