Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006961.3 | NP_008892.2 | 6 | 789 | ||
GenBank mRNA | AK055567.1 | - | 5 | 670 | ||
AK057330.1 | - | 5 | 964 | |||
AK292260.1 | - | 6 | 987 | |||
AL834237.1 | - | 1 | 163 | |||
BC047091.1 | - | 6 | 776 | 5270853 |