Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115873
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
24
3239
NM_001178107.1
NP_001171578.1
23
3152
NM_001178108.1
NP_001171579.1
23
3146
NM_001178109.1
NP_001171580.1
22
3056
NM_001178110.1
NP_001171581.1
21
2966
NM_001178113.1
NP_001171584.1
15
2696
NM_001178115.1
NP_001171586.1
9
2047
NM_007150.3
NP_009081.2
23
3143
XM_005274730.2
XP_005274787.1
25
3346
XM_005274731.2
XP_005274788.1
25
3343
XM_005274735.2
XP_005274792.1
24
3256
XM_005274737.2
XP_005274794.1
24
3250
XM_005274738.2
XP_005274795.1
24
3247
XM_005274740.2
XP_005274797.1
23
3160
XM_005274741.2
XP_005274798.1
23
3160
XM_005274742.2
XP_005274799.1
23
3157
XM_005274744.2
XP_005274801.1
22
3073
XM_005274745.2
XP_005274802.1
22
3070
XM_005274746.3
XP_005274803.1
21
2989
XM_011531194.1
XP_011529496.1
23
3151
XM_011531195.1
XP_011529497.1
24
3148
XM_017029821.1
XP_016885310.1
26
3478
XM_017029822.1
XP_016885311.1
27
3498
XM_017029823.1
XP_016885312.1
26
3475
XM_017029824.1
XP_016885313.1
25
3394
XM_017029825.1
XP_016885314.1
25
3391
XM_017029826.1
XP_016885315.1
25
3388
XM_017029827.1
XP_016885316.1
25
3385
XM_017029828.1
XP_016885317.1
25
3382
XM_017029829.1
XP_016885318.1
24
3301
XM_017029830.1
XP_016885319.1
24
3295
XM_017029831.1
XP_016885320.1
24
3292
XM_017029832.1
XP_016885321.1
24
3283
XM_017029833.1
XP_016885322.1
23
3217
XM_017029834.1
XP_016885323.1
23
3205
XM_017029835.1
XP_016885324.1
23
3166
GenBank mRNA
AK056517.1
-
12
2202
AK095258.1
-
9
2019
AY997296.1
-
23
3132
BX537525.1
-
9
3721
BX640940.1
-
15
2443
Y09538.1
-
15
2423
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0