Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159296.2 | NP_001152768.1 | 4 | 872 | ||
NM_001324099.1 | NP_001311028.1 | 2 | 408 | |||
NM_001324100.1 | NP_001311029.1 | 3 | 482 | |||
NM_001324102.1 | NP_001311031.1 | 4 | 882 | |||
NM_001324104.1 | NP_001311033.1 | 4 | 648 | |||
NM_001324105.1 | NP_001311034.1 | 3 | 976 | |||
NM_007250.5 | NP_009181.2 | 3 | 1284 | |||
NR_136704.1 | 3 | 482 | ||||
NR_136705.1 | 3 | 1284 | ||||
XM_005261977.2 | XP_005262034.1 | 4 | 934 | |||
XM_005261979.3 | XP_005262036.1 | 4 | 617 | |||
XM_006724575.2 | XP_006724638.1 | 3 | 1284 | |||
XM_006724576.3 | XP_006724639.1 | 3 | 1284 | |||
XM_011530759.2 | XP_011529061.1 | 2 | 1210 | |||
XM_017029250.1 | XP_016884739.1 | 3 | 861 | |||
GenBank mRNA | AB209004.1 | - | 3 | 541 | ||
AK296156.1 | - | 4 | 796 | |||
BC031355.1 | - | 3 | 451 | 4800675 | ||
BC105130.1 | - | 3 | 593 | 8144133 | ||
BC112109.1 | - | 3 | 593 | 8327577 | ||
BX641066.1 | - | 4 | 534 | |||
HF546207.1 | - | 3 | 428 | |||
HF546208.1 | - | 2 | 354 | |||
HF546209.1 | - | 3 | 428 |