Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164391.1 | NP_001157863.1 | 3 | 347 | ||
NM_001164392.1 | NP_001157864.1 | 2 | 427 | |||
NM_007358.3 | NP_031384.1 | 2 | 427 | |||
XM_011541016.1 | XP_011539318.1 | 2 | 379 | |||
XM_011541017.2 | XP_011539319.1 | 2 | 443 | |||
XM_011541019.1 | XP_011539321.1 | 3 | 299 | |||
XM_017000676.1 | XP_016856165.1 | 2 | 282 | |||
XM_017000677.1 | XP_016856166.1 | 2 | 379 | |||
XM_017000678.1 | XP_016856167.1 | 3 | 336 | |||
XM_017000679.1 | XP_016856168.1 | 2 | 379 | |||
GenBank mRNA | AF072814.1 | - | 1 | 121 | ||
AF086908.1 | - | 2 | 352 | |||
AJ010014.1 | - | 2 | 378 | |||
AK290318.1 | - | 2 | 379 | |||
AK302776.1 | - | 3 | 286 | |||
BC001897.1 | - | 2 | 346 | |||
BC010013.2 | - | 2 | 351 | 4308565 | ||
DB173315.1 | - | 2 | 427 |