Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308142.1 | NP_001295071.1 | 10, 11 | 1268 | ||
NM_014048.4 | NP_054767.3 | 10, 11 | 1269 | |||
XM_005255452.3 | XP_005255509.1 | 11, 12 | 1423 | |||
XM_005255453.4 | XP_005255510.1 | 10, 11 | 1907 | |||
XM_005255455.3 | XP_005255512.1 | 8, 9 | 1253 | |||
XM_006720908.3 | XP_006720971.1 | 11, 12 | 2069 | |||
XM_006720909.3 | XP_006720972.1 | 11, 12 | 1398 | |||
XM_006720912.3 | XP_006720975.1 | 8, 9 | 3208 | |||
XM_006720913.2 | XP_006720976.1 | 11, 12 | 1423 | |||
XM_006720914.2 | XP_006720977.1 | 11, 12 | 1423 | |||
XM_011522568.2 | XP_011520870.1 | 10, 11 | 1238 | |||
XM_011522569.2 | XP_011520871.1 | 8, 9 | 1035 | |||
XM_017023501.1 | XP_016878990.1 | 10, 11 | 1234 | |||
XM_017023502.1 | XP_016878991.1 | 10, 11 | 1291 | |||
XM_017023503.1 | XP_016878992.1 | 9, 10 | 4436 | |||
XM_017023504.1 | XP_016878993.1 | 8, 9 | 1253 | |||
XM_017023505.1 | XP_016878994.1 | 8, 9 | 3208 | |||
GenBank mRNA | AB033069.2 | - | 2, 3 | 275 | ||
AK294765.1 | - | 10, 11 | 1260 | |||
BC034934.1 | - | 9, 10 | 1221 | |||
BC136260.1 | - | 8, 9 | 1113 | 9020247 | ||
BC171750.1 | - | 8, 9 | 1113 | 9053251 |