Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014252.3 | NP_055067.1 | 7 | 3119 | ||
GenBank mRNA | AF112968.1 | - | 8 | 1755 |