Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002470.3 | NP_002461.2 | 3 | 273 | ||
XM_011523870.2 | XP_011522172.1 | 3 | 1640 | |||
XM_011523871.2 | XP_011522173.1 | 3 | 273 | |||
GenBank mRNA | BP232245.1 | - | 3 | 273 | ||
BP233220.1 | - | 3 | 273 | |||
BQ956249.1 | - | 2 | 265 | |||
X13988.1 | - | 2 | 269 |