Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303241.1 | NP_001290170.1 | 10 | 1194 | ||
XM_005265953.1 | XP_005266010.1 | 9 | 1194 | |||
XM_011534605.2 | XP_011532907.1 | 8 | 1087 | |||
XM_011534606.1 | XP_011532908.1 | 8 | 1102 | |||
XM_011534607.1 | XP_011532909.1 | 9 | 1297 | |||
XM_017009670.1 | XP_016865159.1 | 7 | 979 | |||
XM_017009671.1 | XP_016865160.1 | 7 | 641 | |||
XM_017009672.1 | XP_016865161.1 | 8 | 1102 | |||
GenBank mRNA | AB033020.1 | - | 10 | 1160 | ||
BC152469.1 | - | 10 | 1160 | 8862606 |