Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF2 SIRNA Assays ›
Gene Symbol:
IKZF2
Gene Name:
IKAROS family zinc finger 2
Gene Aliases:
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location:
Chr.2: 212999684 - 213152753 on Build GRCh38
UniGene ID:
Hs.604950
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116127
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079526.1
NP_001072994.1
7
829
NM_016260.2
NP_057344.2
6
760
XM_005246384.4
XP_005246441.1
6
870
XM_005246385.3
XP_005246442.1
6
900
XM_005246386.4
XP_005246443.1
6
751
XM_011510802.2
XP_011509104.1
7
942
XM_011510803.2
XP_011509105.1
7
791
XM_011510804.2
XP_011509106.1
7
714
XM_011510805.2
XP_011509107.1
7
803
XM_011510807.2
XP_011509109.1
7
978
XM_011510808.2
XP_011509110.1
7
828
XM_011510809.2
XP_011509111.1
7
840
XM_011510810.2
XP_011509112.1
7
754
XM_011510811.2
XP_011509113.1
6
875
XM_011510812.2
XP_011509114.1
6
736
XM_011510814.2
XP_011509116.1
6
1234
XM_011510815.2
XP_011509117.1
6
682
XM_011510816.2
XP_011509118.1
4
683
XM_011510817.2
XP_011509119.1
6
762
XM_011510818.2
XP_011509120.1
4
6337
XM_011510819.2
XP_011509121.1
6
751
XM_017003589.1
XP_016859078.1
6
822
XM_017003590.1
XP_016859079.1
6
792
XM_017003591.1
XP_016859080.1
4
1225
XM_017003592.1
XP_016859081.1
6
683
GenBank mRNA
AF130863.1
-
6
760
AK301503.1
-
6
756
AY587062.1
-
7
744
AY587063.1
-
7
706
AY587064.1
-
5
591
AY587065.1
-
6
597
AY587066.1
-
5
513
AY587067.1
-
5
408
AY587068.1
-
5
591
AY587069.1
-
5
513
AY587070.1
-
3
156
AY587073.1
-
6
770
BC028936.1
-
7
829
4540576
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
biological_process
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
protein homodimerization activity
transcription regulatory region DNA binding
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0