Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193285.2 | NP_001180214.1 | 4 | 472 | ||
NM_001321058.1 | NP_001307987.1 | 3 | 344 | |||
NM_001321059.1 | NP_001307988.1 | 3 | 289 | |||
NM_001321060.1 | NP_001307989.1 | 4 | 472 | |||
NM_001321061.1 | NP_001307990.1 | 3 | 344 | |||
NM_001321062.1 | NP_001307991.1 | 2 | 161 | |||
NM_001321063.1 | NP_001307992.1 | 3 | 289 | |||
NM_001321064.1 | NP_001307993.1 | 4 | 437 | |||
NM_016539.3 | NP_057623.2 | 4 | 472 | |||
GenBank mRNA | AF233396.1 | - | 4 | 467 | ||
AK074810.1 | - | 4 | 435 | |||
AK225567.1 | - | 4 | 440 | |||
AK293348.1 | - | 3 | 305 | |||
AK299420.1 | - | 3 | 287 | |||
AK301247.1 | - | 3 | 252 | |||
AK315048.1 | - | 4 | 436 | |||
AL559383.3 | - | 4 | 410 | |||
BC004218.2 | - | 3 | 284 | 3352203 | ||
BC005026.1 | - | 4 | 410 | 2820472 | ||
BC028220.1 | - | 4 | 418 | 5217599 | ||
BE281103.1 | - | 3 | 270 | |||
BP418952.1 | - | 2 | 106 | |||
BQ060703.1 | - | 3 | 185 | |||
CN364175.1 | - | 4 | 415 | |||
CN364177.1 | - | 4 | 475 | |||
CR457200.1 | - | 4 | 407 |