Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193285.2 | NP_001180214.1 | 7 | 1351 | ||
NM_001321058.1 | NP_001307987.1 | 7 | 1304 | |||
NM_001321059.1 | NP_001307988.1 | 7 | 1249 | |||
NM_001321060.1 | NP_001307989.1 | 8 | 1392 | |||
NM_001321061.1 | NP_001307990.1 | 6 | 1223 | |||
NM_001321062.1 | NP_001307991.1 | 6 | 1121 | |||
NM_001321063.1 | NP_001307992.1 | 7 | 1209 | |||
NM_001321064.1 | NP_001307993.1 | 8 | 1357 | |||
NM_016539.3 | NP_057623.2 | 8 | 1432 | |||
GenBank mRNA | AF233396.1 | - | 8 | 1427 | ||
AK074810.1 | - | 8 | 1395 | |||
AK225567.1 | - | 8 | 1400 | |||
AK293348.1 | - | 7 | 1265 | |||
BC004218.2 | - | 6 | 1163 | 3352203 | ||
BC005026.1 | - | 8 | 1370 | 2820472 | ||
BC028220.1 | - | 7 | 1297 | 5217599 |