Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128833.1 | NP_001122305.1 | 3 | 733 | ||
NM_020899.3 | NP_065950.2 | 3 | 809 | |||
XM_006721563.3 | XP_006721626.1 | 3 | 660 | |||
XM_006721564.2 | XP_006721627.1 | 3 | 640 | |||
XM_011523972.2 | XP_011522274.1 | 3 | 1092 | |||
GenBank mRNA | AB040971.2 | - | 3 | 733 | ||
AK122826.1 | - | 1 | 61 | |||
AK122971.1 | - | 3 | 633 | |||
AY302699.1 | - | 3 | 649 | |||
BC043352.2 | - | 3 | 758 | 6175382 |