Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBZF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039618.2 | NP_001034707.1 | 1 | 3623 | ||
NR_028024.1 | 2 | 2557 | ||||
NR_028025.1 | 3 | 2344 | ||||
XM_006718642.3 | XP_006718705.1 | 3 | 3190 | |||
XM_011545195.2 | XP_011543497.1 | 2 | 3403 | |||
XM_017018086.1 | XP_016873575.1 | 4 | 2469 | |||
XM_017018088.1 | XP_016873577.1 | 2 | 2912 | |||
XM_017018090.1 | XP_016873579.1 | 3 | 2682 | |||
GenBank mRNA | AF039942.1 | - | 2 | 2505 | ||
BC060807.1 | - | 1 | 3615 | 30345789 | ||
DQ128105.1 | - | 2 | 2075 |