Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RFWD2 SIRNA Assays ›
Gene Symbol:
RFWD2
Gene Name:
ring finger and WD repeat domain 2
Gene Aliases:
COP1, RNF200
Chromosome Location:
Chr.1: 175944826 - 176207279 on Build GRCh38
UniGene ID:
Hs.523744
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116337
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RFWD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001740.3
NP_001001740.1
4
885
NM_022457.6
NP_071902.2
4
885
XM_005245447.2
XP_005245504.1
4
920
XM_005245448.2
XP_005245505.1
4
920
XM_006711487.2
XP_006711550.1
4
920
XM_011509881.1
XP_011508183.1
4
920
XM_017002059.1
XP_016857548.1
4
920
XM_017002060.1
XP_016857549.1
4
920
XM_017002061.1
XP_016857550.1
4
920
XM_017002062.1
XP_016857551.1
4
920
XM_017002063.1
XP_016857552.1
4
920
XM_017002064.1
XP_016857553.1
4
920
XM_017002065.1
XP_016857554.1
4
920
XM_017002066.1
XP_016857555.1
4
920
XM_017002067.1
XP_016857556.1
4
920
XM_017002068.1
XP_016857557.1
4
920
XM_017002070.1
XP_016857559.1
3
626
XM_017002076.1
XP_016857565.1
2
84
XM_017002078.1
XP_016857567.1
4
274
XM_017002079.1
XP_016857568.1
4
701
GenBank mRNA
AF508940.1
-
4
619
AF527539.1
-
4
882
AK001278.1
-
3
184
AK314750.1
-
4
817
AY509921.1
-
4
619
BC020845.1
-
4
792
BC094728.1
-
4
873
5271629
KJ534928.1
-
4
619
KJ534929.1
-
4
619
KJ534930.1
-
4
619
KJ534931.1
-
4
619
KJ534932.1
-
4
619
KJ535076.1
-
3
521
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006250116, XM_006250114, XM_006250113, XM_006250115.This siRNA also targets the following transcripts in Mouse: NM_011931, AF151110, BC158002, BC082804.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
response to ionizing radiation
protein ubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0