Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116410
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
6
1113
NM_001174093.1
NP_001167564.1
5
773
NM_001174094.1
NP_001167565.1
6
1110
NM_001174095.1
NP_001167566.1
5
632
NM_001174096.1
NP_001167567.1
6
836
NM_001323638.1
NP_001310567.1
10
1483
NM_001323641.1
NP_001310570.1
6
1063
NM_001323642.1
NP_001310571.1
8
1365
NM_001323643.1
NP_001310572.1
8
1143
NM_001323644.1
NP_001310573.1
5
862
NM_001323645.1
NP_001310574.1
7
1206
NM_001323646.1
NP_001310575.1
9
1444
NM_001323647.1
NP_001310576.1
7
976
NM_001323648.1
NP_001310577.1
6
916
NM_001323649.1
NP_001310578.1
7
979
NM_001323650.1
NP_001310579.1
9
1211
NM_001323651.1
NP_001310580.1
7
879
NM_001323652.1
NP_001310581.1
8
1028
NM_001323653.1
NP_001310582.1
8
1135
NM_001323654.1
NP_001310583.1
8
1022
NM_001323655.1
NP_001310584.1
8
1132
NM_001323656.1
NP_001310585.1
6
807
NM_001323657.1
NP_001310586.1
6
810
NM_001323658.1
NP_001310587.1
6
649
NM_001323659.1
NP_001310588.1
6
867
NM_001323660.1
NP_001310589.1
6
864
NM_001323661.1
NP_001310590.1
7
1010
NM_001323662.1
NP_001310591.1
8
1412
NM_001323663.1
NP_001310592.1
9
1351
NM_001323664.1
NP_001310593.1
8
1335
NM_001323665.1
NP_001310594.1
7
1253
NM_001323666.1
NP_001310595.1
6
1193
NM_001323671.1
NP_001310600.1
7
1089
NM_001323672.1
NP_001310601.1
8
1141
NM_001323673.1
NP_001310602.1
7
1086
NM_001323674.1
NP_001310603.1
4
611
NM_001323675.1
NP_001310604.1
4
721
NM_001323676.1
NP_001310605.1
6
946
NM_001323677.1
NP_001310606.1
6
943
NM_001323678.1
NP_001310607.1
4
888
NM_030751.5
NP_110378.3
6
833
XM_006717498.2
XP_006717561.1
9
1271
XM_011519643.2
XP_011517945.1
9
1574
XM_017016597.1
XP_016872086.1
9
1173
XM_017016598.1
XP_016872087.1
9
4303
XM_017016599.1
XP_016872088.1
8
4141
XM_017016600.1
XP_016872089.1
10
4382
XM_017016601.1
XP_016872090.1
9
4300
XM_017016602.1
XP_016872091.1
10
4379
XM_017016603.1
XP_016872092.1
9
4217
GenBank mRNA
AK091478.1
-
5
733
AK294775.1
-
6
1028
AK296244.1
-
6
756
AK299541.1
-
3
370
AK299666.1
-
6
867
AK300830.1
-
5
580
AK307510.1
-
7
839
AK308822.1
-
8
978
AK309399.1
-
7
951
AK310924.1
-
7
940
AK311116.1
-
6
846
AK311137.1
-
4
572
AK314683.1
-
6
794
AL831979.1
-
8
1091
BC009572.1
-
6
789
BC107781.1
-
6
789
BC112392.1
-
6
789
40036600
BX647794.1
-
10
1484
U12170.1
-
6
794
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0