Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168378.1 | NP_001161850.1 | 3 | 511 | ||
NM_001168379.1 | NP_001161851.1 | 3 | 338 | |||
NM_032153.5 | NP_115529.2 | 3 | 637 | |||
XM_017007314.1 | XP_016862803.1 | 3 | 3120 | |||
GenBank mRNA | AF332509.1 | - | 1 | 1030 | ||
AK056262.1 | - | 3 | 448 | |||
AK056394.1 | - | 1 | 822 | |||
AK094286.1 | - | 1 | 134 | |||
AK293981.1 | - | 3 | 338 | |||
AK294831.1 | - | 3 | 511 | |||
BC029507.1 | - | 3 | 416 | |||
BC126267.1 | - | 3 | 273 | 8991983 | ||
BC136339.1 | - | 3 | 273 | 9020326 | ||
DA066220.1 | - | 3 | 366 |