Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007078.1 | - | 5 | 1102 | ||
BC008386.1 | - | 4 | 1063 | |||
BC019234.1 | - | 5 | 1106 | |||
BC108740.1 | - | 4 | 820 |