Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002567.3 | NP_002558.1 | 1, 2 | 469 | ||
GenBank mRNA | AK226006.1 | - | 1, 2 | 248 | ||
AK299414.1 | - | 1, 2 | 228 | |||
AK308056.1 | - | 1, 2 | 227 | |||
AK311927.1 | - | 1, 2 | 240 | |||
BC008714.2 | - | 1, 2 | 242 | 2822606 | ||
BC017396.1 | - | 1, 2 | 263 | 4074533 | ||
BC018799.1 | - | 1, 2 | 233 | |||
BC031102.1 | - | 1, 2 | 231 | 5300366 | ||
BF978793.1 | - | 1, 2 | 311 | |||
BP347345.1 | - | 1, 2 | 494 | |||
D16111.1 | - | 1, 2 | 234 | |||
EU794643.1 | - | 1, 2 | 233 | |||
HM131449.1 | - | 1, 2 | 225 | |||
X75252.1 | - | 1, 2 | 253 | |||
X85033.1 | - | 1, 2 | 146 |