Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005992.1 | NP_005983.1 | 5 | 761 | ||
NM_080646.1 | NP_542377.1 | 5 | 761 | |||
NM_080647.1 | NP_542378.1 | 5 | 761 | |||
XM_006724312.2 | XP_006724375.1 | 5 | 930 | |||
XM_017028925.1 | XP_016884414.1 | 4 | 806 | |||
XM_017028926.1 | XP_016884415.1 | 5 | 896 | |||
XM_017028928.1 | XP_016884417.1 | 4 | 805 | |||
GenBank mRNA | AF012130.1 | - | 5 | 761 | ||
AF012131.1 | - | 5 | 761 | |||
AF373867.1 | - | 5 | 761 |