Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278734.1 | NP_001265663.1 | 5 | 801 | ||
NM_016324.3 | NP_057408.2 | 2 | 485 | |||
NM_016325.3 | NP_057409.1 | 4 | 690 | |||
NM_133502.2 | NP_598009.1 | 5 | 786 | |||
XM_011526327.1 | XP_011524629.1 | 4 | 224 | |||
XM_017026174.1 | XP_016881663.1 | 3 | 491 | |||
XM_017026175.1 | XP_016881664.1 | 4 | 759 | |||
GenBank mRNA | AB029149.1 | - | 2 | 445 | ||
AB029150.1 | - | 4 | 592 | |||
AF275680.1 | - | 5 | 465 | |||
AK222855.1 | - | 2 | 413 | |||
AK222878.1 | - | 2 | 413 | |||
AK295036.1 | - | 2 | 365 | |||
AK296929.1 | - | 5 | 375 | |||
AK297466.1 | - | 5 | 376 | |||
AK307906.1 | - | 5 | 363 | |||
AK316429.1 | - | 5 | 712 | |||
BC009763.2 | - | 4 | 313 | 2966764 | ||
BT007304.1 | - | 4 | 287 | |||
DA050458.1 | - | 4 | 270 | |||
DC362701.1 | - | 5 | 366 |