Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000338.2 | NP_000329.2 | 22 | 2949 | ||
NM_001184832.1 | NP_001171761.1 | 22 | 2949 | |||
XM_005254606.1 | XP_005254663.1 | 22 | 2949 | |||
GenBank mRNA | BX647067.1 | - | 17 | 6873 | ||
BX647484.1 | - | 18 | 6856 | |||
EF559316.2 | - | 21 | 2747 | |||
U58130.1 | - | 21 | 2752 |