Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000446.5 | NP_000437.3 | 9 | 1167 | ||
GenBank mRNA | BC074719.2 | - | 9 | 1099 | 30915169 | |
D84371.1 | - | 9 | 1124 | |||
M63012.1 | - | 9 | 1079 | |||
M63013.1 | - | 9 | 1018 | |||
M63014.1 | - | 8 | 984 | |||
S64615.1 | - | 9 | 1079 | |||
U53784.1 | - | 9 | 1124 | |||
Z70723.1 | - | 9 | 1124 |