Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001697.2 | NP_001688.1 | 6 | 553 | ||
GenBank mRNA | AF088071.1 | - | 4 | 348 | ||
AK093795.1 | - | 2 | 1474 | |||
AK124187.1 | - | 5 | 2588 | |||
AK222608.1 | - | 6 | 485 | |||
AK222962.1 | - | 6 | 480 | |||
AK311796.1 | - | 6 | 480 | |||
BC021233.2 | - | 6 | 466 | 4126036 | ||
BC022865.1 | - | 6 | 484 | 4103001 | ||
BT019836.1 | - | 6 | 462 | |||
CD174837.1 | - | 6 | 523 | |||
CR456822.1 | - | 6 | 462 | |||
X83218.1 | - | 6 | 498 |