Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161587.1 | NP_001155059.1 | 7 | 1165 | ||
NM_002103.4 | NP_002094.2 | 8 | 1357 | |||
NR_027763.1 | 7 | 1175 | ||||
GenBank mRNA | AK223579.1 | - | 8 | 1313 | ||
AK291672.1 | - | 8 | 1313 | |||
AK294349.1 | - | 3 | 273 | |||
AK300369.1 | - | 8 | 1145 | |||
AK302738.1 | - | 7 | 1131 | |||
AK303712.1 | - | 6 | 1013 | |||
BC002617.2 | - | 8 | 1296 | 3143019 | ||
BC003182.1 | - | 7 | 1122 | 2958297 | ||
BC007688.2 | - | 8 | 1283 | 3953692 | ||
J04501.1 | - | 8 | 1276 | |||
U32573.1 | - | 8 | 1258 |