Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171993.1 | NP_001165464.1 | 14 | 1405 | ||
NM_002150.2 | NP_002141.1 | 12 | 929 | |||
GenBank mRNA | AK057510.1 | - | 14 | 1405 | ||
AK290826.1 | - | 12 | 925 | |||
BC024287.1 | - | 12 | 911 | 5087393 | ||
X72389.1 | - | 12 | 914 |