Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018425.3 | NP_060895.1 | 1, 2 | 502 | ||
GenBank mRNA | AJ303098.1 | - | 1, 2 | 502 | ||
BC003167.1 | - | 1, 2 | 474 | 2905670 | ||
BT007330.1 | - | 1, 2 | 445 |