Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003241.3 | NP_003232.2 | 11 | 1480 | ||
XM_011534042.1 | XP_011532344.1 | 12 | 1629 | |||
GenBank mRNA | AK300523.1 | - | 11 | 1349 | ||
AK312936.1 | - | 11 | 1478 | |||
BC007003.1 | - | 11 | 1475 | 3950865 | ||
L34840.1 | - | 11 | 1451 | |||
U31905.1 | - | 11 | 1451 |