Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
117177
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
19
2902
NM_001258380.1
NP_001245309.1
19
2664
NM_001321103.1
NP_001308032.1
20
3286
NM_001321104.1
NP_001308033.1
20
3247
NM_001321105.1
NP_001308034.1
20
3247
NM_001321106.1
NP_001308035.1
20
3235
NM_001321107.1
NP_001308036.1
19
2902
NM_001321108.1
NP_001308037.1
19
2914
NM_003615.4
NP_003606.3
20
3036
NR_135541.1
19
3060
NR_135542.1
18
2727
NR_135543.1
18
2727
NR_135544.1
19
3060
XM_005265598.4
XP_005265655.1
19
3168
XM_005265600.4
XP_005265657.1
19
3130
XM_006713421.2
XP_006713484.1
20
3274
XM_011534256.2
XP_011532558.1
20
3547
XM_011534258.1
XP_011532560.1
20
3250
XM_011534261.2
XP_011532563.1
20
3512
XM_011534263.2
XP_011532565.1
19
2839
XM_011534265.1
XP_011532567.1
19
2878
XM_017007526.1
XP_016863015.1
21
3171
XM_017007527.1
XP_016863016.1
19
2863
XM_017007528.1
XP_016863017.1
13
1787
XM_017007529.1
XP_016863018.1
13
1769
GenBank mRNA
AB012130.1
-
16
2350
AF047033.1
-
20
3036
AF053755.1
-
16
2342
AF089726.1
-
19
2664
CR627428.1
-
12
1618
EU499349.1
-
19
2829
EU934246.1
-
20
3162
EU934247.1
-
20
3174
EU934248.1
-
20
3213
EU934249.1
-
19
2829
EU934250.1
-
20
3174
FJ178574.1
-
19
2987
FJ178575.1
-
18
2654
FJ178576.1
-
18
2654
GU354307.1
-
19
2987
GU354308.1
-
19
2841
GU354309.1
-
18
2666
GU354310.1
-
18
2666
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0