Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159629.1 | NP_001153101.1 | 2 | 730 | ||
NM_003645.3 | NP_003636.2 | 2 | 730 | |||
GenBank mRNA | AF096290.1 | - | 2 | 702 | ||
AK123905.1 | - | 2 | 124 | |||
AK222859.1 | - | 2 | 701 | |||
AK223145.1 | - | 2 | 699 | |||
AK290262.1 | - | 2 | 730 | |||
BC057770.1 | - | 2 | 693 | 30348317 | ||
D88308.1 | - | 2 | 720 |