Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079855.1 | NP_001073324.1 | 4 | 455 | ||
NM_001184702.1 | NP_001171631.1 | 4 | 455 | |||
NM_001184703.1 | NP_001171632.1 | 5 | 548 | |||
NM_001184704.1 | NP_001171633.1 | 3 | 313 | |||
NM_003918.2 | NP_003909.2 | 5 | 548 | |||
XM_011545600.2 | XP_011543902.1 | 4 | 386 | |||
XM_017029927.1 | XP_016885416.1 | 5 | 498 | |||
XM_017029928.1 | XP_016885417.1 | 5 | 473 | |||
XM_017029929.1 | XP_016885418.1 | 5 | 658 | |||
XM_017029930.1 | XP_016885419.1 | 4 | 363 | |||
XM_017029931.1 | XP_016885420.1 | 4 | 386 | |||
GenBank mRNA | AK098046.1 | - | 3 | 221 | ||
AK292496.1 | - | 4 | 363 | |||
BC023152.1 | - | 4 | 322 | 3923041 | ||
DQ424901.1 | - | 3 | 266 | |||
U94362.1 | - | 5 | 549 | |||
U94363.1 | - | 4 | 329 |