Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002522.3 | NP_002513.2 | 3 | 892 | ||
GenBank mRNA | AK124701.1 | - | 1 | 1335 | ||
AK127335.1 | - | 3 | 892 | |||
BC089441.1 | - | 3 | 876 | 30555728 | ||
BX537550.1 | - | 1 | 1431 | |||
DA188847.1 | - | 1 | 23 | |||
U61849.1 | - | 3 | 866 |