Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272087.1 | NP_001259016.1 | 5 | 1655 | ||
NM_001272088.1 | NP_001259017.1 | 3 | 483 | |||
NM_005071.2 | NP_005062.1 | 2 | 261 | |||
NR_073589.1 | 3 | 419 | ||||
XM_006722842.1 | XP_006722905.1 | 3 | 444 | |||
XM_006722844.1 | XP_006722907.1 | 3 | 337 | |||
XM_017027152.1 | XP_016882641.1 | 3 | 532 | |||
GenBank mRNA | AB209199.1 | - | 2 | 3422 | ||
AK302380.1 | - | 3 | 396 | |||
AK312996.1 | - | 3 | 527 | |||
BC028721.2 | - | 3 | 405 | 5269300 | ||
BC040604.1 | - | 5 | 1610 | 5270965 | ||
BI829415.1 | - | 3 | 371 | |||
DB027852.1 | - | 3 | 419 | |||
DR004792.1 | - | 3 | 469 | |||
U18244.1 | - | 2 | 261 |